Order Entry
Belgium
ContactUsLinkComponent
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Catalog # 1.19814.0500
Supplier:  Merck
CAS Number:  
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Catalog # 1.19814.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Fluoride standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    NaF in H₂O

Specifications

About this item