Order Entry
Belgium
ContactUsLinkComponent
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Catalog # MOLE21833767-500G
Supplier:  Molekula
CAS Number:  
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Catalog # MOLE21833767-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :132.16 g/mol
Merck Index:13,05233
Boiling Pt:188 °C (1013 hPa)
Density:1.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-888-7
CAS Number:100-79-8
Formula:C₆H₁₂O₃
MDL Number:MFCD00063238