(±)-2-Methylenanthic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1188-02-9
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00142944
Boiling Pt:85…87 °C (0.5 mmHg)
Density:0.9122 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:144.21 g/mol
EINECS:214-704-2
:GHS07
Specification Test Results
- Molar Mass:144.22 g/mol
- Boiling Point:140 °C (40 hPa)
- Flash Point:110 °C
- Density:0.92 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together