Order Entry
Belgium
Orders LinkContactUsLinkComponent
Barium chloride 10% in aqueous solution

Barium chloride 10% in aqueous solution

5 L
Catalog # FFCHFF20705000
Irritant
Supplier:   FF CHEMICALS
Barium chloride 10% in aqueous solution
Catalog # FFCHFF20705000
Supplier:  FF CHEMICALS
CAS Number:  10361-37-2
Barium chloride 10% in aqueous solution

5 L

CAS Number:  10361-37-2
Price
Inquire For Price
Log in for availability
1 * 5 L
Product Details & Documents
  • CAS Number:10361-37-2
  • Formula:BaCl₂
  • MDL Number:MFCD00003445
  • Molecular Weight:208.24 g/mol
  • Boiling Pt:∼100 °C (1013 hPa)
  • Storage Temperature:Ambient
  • EINECS:233-788-1
  • UN:3287
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.