Order Entry
Belgium
ContactUsLinkComponent
 
 
Potassium hydroxide 45% in aqueous solution
  LOVI2418634
 :  Lovibond
 :  
 
Potassium hydroxide 45% in aqueous solution
  LOVI2418634
 :  Lovibond
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

 

 

 :
 :56.11 g/mol
 :Ambient
 :215-181-3
 :1814
 :8,II
 :GHS05
 :1310-58-3
 :KOH
 :MFCD00003553