Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
 
undefined
 
 
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
 

 

  • Assay (Active chlorine)(at filling):
    Min. 11,5 %
  • Alkalinity:
    Max. 0.5 meq/g
  • Chlororometric degrees (Fr;at packaging):
    min 36,30°
  • Hg (Mercury):
    Max. 10 ppm

 

Frequently Bought Together