Orders LinkContactUsLinkComponent
Multirack™ Jr.
Supplier:   Simport Scientific
Multirack™ Jr.
Supplier:  Simport Scientific
Certificates
Multirack™ Jr.
Multirack™ Jr.
14 Options Available Below
Product Details & Documents
These MultiRack™ Jr. are ideal for use in incubators, refrigerators, freezers, under lab hoods and om benchtops.
  • Made of acetal
  • Available in three models for 13 mm, 16 mm and 25 to 30 mm tubes
  • Compact, lightweight, stackable and submersible
  • 3-tier facilitates stability of tubes
  • Base has rounded wells with drain holes
A newly designed tube support that can be used all around the lab. It is available in three models to accommodate a full range of laboratory test tubes and centrifuge tubes up to 30 mm in diameter. Although it is one of the most attractive racks available today, it offers all the advantages required by the modern laboratory. Made of highly resistant acetal polymer, it will not shatter or stain in contact with most laboratory chemicals. No coating to worry about, which can chip, peel or rust in a water bath.
The MultiRack™ is made of three tiers to facilitate the insertion and stability of tubes. The base tier has rounded wells with drain holes. Convenient handles on each side of the rack will ensure a safe grip when carrying. Interlocking feet allow for stacking.
A great acquisition when space is more limited.
 : Available in five attractive colours.
Product Family Options
MultiRack™ Jr., for upto 13 mm tubes, 42 places
Product Information
  • Colour
    No. of Holes
    For tubes
    Availability
    Price
  • Blue
    42
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 174,00 €
    174,00 €
    1 * 10 ST
       174,00 €
  • Green
    42
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 174,00 €
    174,00 €
    1 * 10 ST
       174,00 €
  • Lilac
    42
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 174,00 €
    174,00 €
    1 * 10 ST
       174,00 €
  • Orange
    42
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 174,00 €
    174,00 €
    1 * 10 ST
       174,00 €
  • Yellow
    42
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 174,00 €
    174,00 €
    1 * 10 ST
       174,00 €
MultiRack™ Jr., for upto 16 mm tubes, 30 places
Product Information
  • Colour
    No. of Holes
    For tubes
    Availability
    Price
  • Green
    30
    16 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Lilac
    30
    16 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Orange
    30
    16 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Yellow
    30
    16 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
MultiRack™ Jr., for 25 - 30 mm tubes, 9 places
Product Information
  • Colour
    No. of Holes
    For tubes
    Availability
    Price
  • Blue
    9
    25 - 30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Green
    9
    25 - 30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Lilac
    9
    25 - 30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Orange
    9
    25 - 30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €
  • Yellow
    9
    25 - 30 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 177,00 €
    177,00 €
    1 * 10 ST
       177,00 €