Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenolphthalein 1%, TECHNICAL

Phenolphthalein 1%, TECHNICAL

Glass bottle, 1 L
Catalog # 8626.1000
Flammable
Health
Irritant
Supplier:   VWR Chemicals
Phenolphthalein 1%, TECHNICAL
Catalog # 8626.1000
Supplier:  VWR Chemicals
CAS Number:  77-09-8
Phenolphthalein 1%, TECHNICAL
Phenolphthalein 1%, TECHNICAL

Glass bottle, 1 L

CAS Number:  77-09-8
Price
40,90 €
Log in for availability
1 * 1 L
Product Details & Documents
  • Transition range: pH 8,2-9,8
Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • MDL Number:MFCD00005913
  • Molecular Weight:318.33 g/mol
  • Boiling Pt:82 °C (1013 hPa)
  • Density:0.79 g/cm³ (20 °C)
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:201-004-7
  • UN:1219
  • ADR:3,II
Specifications

Specification Test Results

  • Sensitivity:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.