Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 0.04 mol/l (0.24 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Toxic
Health
Potassium dichromate 0.04 mol/l (0.24 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # 30892.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Potassium dichromate 0.04 mol/l (0.24 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # 30892.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.03992 - 0.04008 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367