ContactUsLinkComponent
Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution, AVS TITRINORM volumetric solution, for COD determination in water
Toxic
Corrosive
Health
Environment
Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution, AVS TITRINORM volumetric solution, for COD determination in water
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Corrosive
Health
Environment
Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution, AVS TITRINORM volumetric solution, for COD determination in water
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.03992 - 0.04008 mol/l
  • NIST traceable:
    Confirmed