Sodium-L(+)-glutamate monohydrate ≥99%, TECHNICAL
Catalog # 27872.298
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99 %
Specification Test Results
About this item
Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate
:
:187.13 g/mol
Merck Index:13,06278
Melting Pt:225…240 °C
Flash Pt:>225 °C
Storage Temperature:Ambient
EINECS:240-313-1
CAS Number:6106-04-3
Formula:C₅H₈NNaO₄·H₂O
MDL Number:MFCD00150138