Order Entry
Belgium
Orders LinkContactUsLinkComponent
Saffron, powder, TECHNICAL

Saffron, powder, TECHNICAL

Glass bottle, 5 g
Catalog # 27481.105
Supplier:   VWR Chemicals
Saffron, powder, TECHNICAL
Catalog # 27481.105
Supplier:  VWR Chemicals
CAS Number:  84604-17-1
Saffron, powder, TECHNICAL
Saffron, powder, TECHNICAL

Glass bottle, 5 g

CAS Number:  84604-17-1
Price
130,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  C.I. 75100
  • CAS Number:84604-17-1
  • Molecular Weight:976.96 g/mol
  • EINECS:283-295-0
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.