trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥99.0%
Catalog # 20284.153
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (calculated on anhydrous):Min. 99.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.1 %
- SO4 (Sulphate):Max. 100 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate
:
:364.35 g/mol
EINECS:236-308-9
:GHS07
Storage Temperature:Ambient
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O
Melting Pt:213…216 °C