Order Entry
Belgium
ContactUsLinkComponent
Dowsil™ / Dow Corning DC4 Electrical Insulation Compound
Dowsil™ / Dow Corning DC4 Electrical Insulation Compound
Catalog # 632227H
Supplier:  DOW CORNING
CAS Number:  
Dowsil™ / Dow Corning DC4 Electrical Insulation Compound
Catalog # 632227H
Supplier:  DOW CORNING
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
Storage Temperature:Ambient