Order Entry
Belgium
ContactUsLinkComponent
emulsion 10% active compound pH 3-4 for use in aqueous, textile, chemical and industrial foam control
emulsion 10% active compound pH 3-4 for use in aqueous, textile, chemical and industrial foam control
Catalog # 632156J
Supplier:  VWR Chemicals
CAS Number:  
undefined
emulsion 10% active compound pH 3-4 for use in aqueous, textile, chemical and industrial foam control
Catalog # 632156J
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient