ContactUsLinkComponent
VWR® ARISTAR® Anion Multi-Component Standard 2 for Ion Chromatography, Avantor
VWR® ARISTAR® Anion Multi-Component Standard 2 for Ion Chromatography, Avantor
Catalog # 458152E
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR® Anion Multi-Component Standard 2 for Ion Chromatography, Avantor
Catalog # 458152E
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Anion multi-component standard 2 for IC
Specifications
About this item
Br 400 ppm   NO3 400 ppm
Cl 200 ppm   PO4 600 ppm
F 100 ppm   SO4 400 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.