Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard 2 in dil. Nitric Acid for ICP-MS
VWR® ARISTAR®, Multi-Element Quality Control Standard 2 in dil. Nitric Acid for ICP-MS
Catalog # 456602B
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard 2 in dil. Nitric Acid for ICP-MS
Catalog # 456602B
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 2 in dil. nitric acid ARISTAR® for ICP-MS

Specifications

About this item

25 Elements in 5% nitric acid solution.

25 Elements 10 mg/L : Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.