Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Calibration Standard 4 in Water with a Trace of Hydrofluoric Acid for ICP-MS
VWR® ARISTAR®, Multi-Element Calibration Standard 4 in Water with a Trace of Hydrofluoric Acid for ICP-MS
Catalog # 456522C
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Calibration Standard 4 in Water with a Trace of Hydrofluoric Acid for ICP-MS
Catalog # 456522C
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Matrix:
    H₂O/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element calibration standard 4, ARISTAR® for ICP-MS

Specifications

About this item

(12 Elements in H₂O with trace of HF ).

12 Elements 10 mg/L : B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.