VWR® ARISTAR®, Tantalum Standard Solution, 1000 mg/L Ta in Water with Hydrofluoric Acid (max. 1%) (from Ta), Standard for ICP
Catalog # 456102J
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O/tr HF
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Tantalum
- Symbol:Ta
Specifications
About this item
Ta in H₂O tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.