Order Entry
Belgium
ContactUsLinkComponent
 
Lecithin (from soy bean)
  298632A
 :  
undefined
Lecithin (from soy bean)
  298632A
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Assay of nitrogen:
    0.6 - 1.2 %
  • Assay (Phosphorus):
    2.0 - 3.3 %
  • Sulfated ash (800°C):
    Max. 10 %
  • Microbiological test:
    Passes test
  • Salmonella species (absent in 10 g):
    Passes test
  • Escherischia coli (absent in 1 g):
    Passes test
  • Enterobacteriaceae (in 1 g):
    Absent

 

 

 :
 :310-129-7
 :8030-76-0
 :MFCD00147406