Order Entry
Belgium
ContactUsLinkComponent
VWR® Sodium Dihydrogen Phosphate Buffer Solution (Sodium dihydrogen orthophosphate/Phosphoric acid) for Chlorine Determination in Water Monitors
VWR® Sodium Dihydrogen Phosphate Buffer Solution (Sodium dihydrogen orthophosphate/Phosphoric acid) for Chlorine Determination in Water Monitors
Catalog # 160918E
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Sodium Dihydrogen Phosphate Buffer Solution (Sodium dihydrogen orthophosphate/Phosphoric acid) for Chlorine Determination in Water Monitors
Catalog # 160918E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • pH buffer type:
    Phosphate
  • Pk:
    25 L
  • Colour:
    Neutral
  • Description:
    Sodium dihydrogen phosphate buffer solution
  • pH accuracy:
    ±0,05
  • pH value:
    2,30

Specifications

About this item