VWR® Lens Cleaning Tissues, Grade 541
Suitable for cleaning various optical lenses such as objectives, binoculars, magnifying glasses, spectacles, as well as optical fibre filaments.
Thickness: 0,04 mm
Weight: 12 g/m²
Frequently Bought Together
Suitable for cleaning various optical lenses such as objectives, binoculars, magnifying glasses, spectacles, as well as optical fibre filaments.
Thickness: 0,04 mm
Weight: 12 g/m²