Counting chambers, Fuchs-Rosenthal
Fournisseur: GLASWARENFABRIK KARL HECHT
Certificats
À propos de cet article
This ruling is used for counting leukocytes, eosinophils and cells in cerebro-spinal fluid.
- Total area of ruling: 16 mm²
- Subdivision: 16 square groups of 1 mm² each
- 16 mini squares (0,25×0,25 mm) each
Frequently Bought Together