Orders LinkContactUsLinkComponent
Counting chambers, Bürker
Counting chambers, Bürker
Counting chambers, Bürker
Certificates
Counting chambers, Bürker
Counting chambers, Bürker
2 Options Available Below
Product Details & Documents
These counting chambers are suitable for blood tests and urine analysis.
  • 169 squares (0,05×0,05 mm) of 0,0025 mm² each for erythrocytes
  • 144 squares (0,2×0,2 mm) of 0,04 mm² each for leucocytes
  • Total area of ruling: 9,3 mm²
Documents
Product Family Options
Product Information
  • Type
    Depth of chamber
    Availability
    Price
  • HECH40443703
    Cell counter with clips, single ruling
    Cell counter with clips, single ruling
    0,10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,70 €
    72,70 €
    1 * 1 ST
       72,70 €
  • 631-0923
    Counting chamber, double ruling
    Counting chamber, double ruling
    Bürker
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,40 €
    45,40 €
    1 * 1 ST
       45,40 €