Filter tips, ART® reload system
Supplier: Thermo Fisher Scientific
Certificates
About this item
This sterile barrier tip reload system combines individually packed blister tubs made of recycled plastic and new hinged racks. The push-down blister tub easily inserts the ART® barrier tips into the hinged racks providing stackable space saving racks that offer easy, one-handed use. Empty hinged racks allow for easy conversion to the ART® Reload system.
- Environmentally friendly packaging
- ART® self-sealing barrier prevent cross-contamination
- Tips are certified DNase-, RNase-, ATP- and pyrogen-free
Specifications
- Catalog No:
- 732-1150
- 732-1143
- 732-1151
- 732-1149
- 732-1152
- 732-1147
- 732-1154
- 732-1153
- 732-1167
- 732-1174
- 732-1144
- 732-1172
- 732-1148
- 732-1178
- 732-1502
- 732-1168
- 732-1176
- 732-1180
- 732-1165
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Empty reload racks
Product Information
Frequently Bought Together