VWR® shiroGEL Mini PAGE Electroblotting System
Product Details & Documents
The shiroGEL Mini PAGE electroblotting system allows quick and easy blotting of three gels simultaneously. Gels fit snugly in blotting cassettes with transfer membranes, which are then placed into the blotting insert. Coiled electrodes within the insert generate a high intensity current, allowing standard transfer to take place in approximately one hour.
- Blots up to three gels simultaneously
- Standard transfer in approximately one hour
Delivery information: Complete system includes buffer tank with power leads and cooling pack; blotting insert; 3 blotting cassettes; 6 fibre pads; 10×10 cm notched plates (2); 10×10 cm plates with spacers (2); and 2×12-tooth, 1 mm thick combs.
Documents
VWR® Products for Electrophoresis
Product Literature
Specifications
- Description:Mini PAGE electroblotting system, shiroGEL, without PAGE components
- Seq:45
- Cat. No.:700-0291
- Capacity:3 gels/transfer
- Tank volume:1200 L
Accessories
Product Information
- Catalog #AvailabilityPrice
- Blotting insert, including 3 cassettes
Product Information
- Catalog #AvailabilityPrice
- Mini cooling pack
Product Information
- Catalog #AvailabilityPrice
- Mini blotting cassette
Product Information
- Catalog #AvailabilityPrice
- Fibre pads for blotting cassette
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



