ContactUsLinkComponent
Pump tubing for AKTA flux
Pump tubing for AKTA flux
Catalog # 29-0609-48
Supplier:  Cytiva
Pump tubing for AKTA flux
Catalog # 29-0609-48
Supplier:  Cytiva
Specifications
  • Description:
    Pump tubing, PFD, size 17
  • Pk:
    1
Specifications
About this item
Pump tubing for AKTA flux, AKTA flux is a versatile cross flow filtration system for sample concentration and diafiltration (buffer exchange) as well as cell harvest and clarification.