Order Entry
Belgium
ContactUsLinkComponent
VWR®, Pure Water viscosity standard
VWR®, Pure Water viscosity standard
Catalog # 99566.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Pure Water viscosity standard
Catalog # 99566.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Standard with certified values at 5 °C, 20 °C, 25 °C & 37 °C
  • Seq:
    10
  • Viscosity at 25 °C:
    1,0018 mPa·s
  • No.:
    VISC-WAT
  • Test method:
    ASTM D2162

Specifications

About this item

Pure Water viscosity standard have certified data for kinematic viscosity, dynamic viscosity and density at 5, 20, 25 and 37 °C.

  • Certified in strict accordance with ASTM D2162 for 20, 25 and 37 °C
  • Density g/ml in accordance with ASTM D1480 for all temperatures
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging

Pure Water Viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

These standards are certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture, for temperatures at 20, 25 and 37 °C. These products have been tested at primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement, thus providing standards that are certified with primary level accuracy.

Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Caution: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.