Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rat Recombinant FGF-basic (Animal free) (from <i>E. coli</i>)

Rat Recombinant FGF-basic (Animal free) (from E. coli)

Rat Recombinant FGF-basic (Animal free) (from <i>E. coli</i>)
Certificates
Rat Recombinant FGF-basic (Animal free) (from <i>E. coli</i>)
Rat Recombinant FGF-basic (Animal free) (from E. coli)

5 Options Available Below

Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases. 

  • High Quality
  • Low Endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT300-02AF-10UG
  • SHBT300-02AF-50UG
  • SHBT300-02AF-100UG
  • SHBT300-02AF-500UG
  • SHBT300-02AF-1MG
  • Protein/peptide name:
  • FGF-basic
  • FGF-basic
  • FGF-basic
  • FGF-basic
  • FGF-basic
  • Protein synonyms:
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Prostatropin|HBGF-2|FGF2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P13109
  • P13109
  • P13109
  • P13109
  • P13109
  • Sequence:
  • MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
  • Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Molecular weight:
  • 16,4 kDa
  • 16,4 kDa
  • 16,4 kDa
  • 16,4 kDa
  • 16,4 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 82,30 €
    82,30 €
    1 * 1 EA
       82,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT300-02AF-10UG
    Protein/peptide name
    FGF-basic
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13109
    Sequence
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 209,00 €
    209,00 €
    1 * 50 µG
       209,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT300-02AF-50UG
    Protein/peptide name
    FGF-basic
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13109
    Sequence
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 374,00 €
    374,00 €
    1 * 1 EA
       374,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT300-02AF-100UG
    Protein/peptide name
    FGF-basic
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13109
    Sequence
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 964,00 €
    964,00 €
    1 * 500 µG
       964,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT300-02AF-500UG
    Protein/peptide name
    FGF-basic
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13109
    Sequence
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 1 680,00 €
    1 680,00 €
    1 * 1 mg
       1 680,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT300-02AF-1MG
    Protein/peptide name
    FGF-basic
    Protein synonyms
    Prostatropin|HBGF-2|FGF2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Rat
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13109
    Sequence
    MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
    Molecular weight
    16,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.