Mouse Recombinant IL-3 (from E. coli)
About this item
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
4 Options Available Below
Product Details & Documents
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Documents
Literature reference - PubMed ID 27786273 Literature reference - PubMed ID 29247574
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-01AF-10UG
- SHBT200-01AF-100UG
- SHBT200-01AF-500UG
- SHBT200-01AF-1MG
- Protein/peptide name:
- IL-3 (AF)
- IL-3 (AF)
- IL-3 (AF)
- IL-3 (AF)
- Protein synonyms:
- MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P01586
- P01586
- P01586
- P01586
- Sequence:
- MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC
- MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC
- MFPTSQVRRG DFTEDTTPNR PVYTTSQVGG LITHVLWEIV EMRKELCNGN SDCMNNDDAL AENNLKLPEI QRNDGCYQTG YNQEICLLKI SSGLLEYHSY LEYMKNNLKD NKKDKARVLQ RDTETLIHIF NQEVKDLHKI VLPTPISNAL LTDKLESQKE WLRTKTIQFI LKSLEEFLKV TLRSTRQT
- MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- Molecular weight:
- 15,2 kDa
- 15,2 kDa
- 15,2 kDa
- 15,2 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-01AF-10UGProtein/peptide nameIL-3 (AF)Protein synonymsMCGF|P-cell stimulating factor|HCGF|Multi-CSFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP01586SequenceMDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVECProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight15,2 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-01AF-100UGProtein/peptide nameIL-3 (AF)Protein synonymsMCGF|P-cell stimulating factor|HCGF|Multi-CSFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP01586SequenceMDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVECProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight15,2 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-01AF-500UGProtein/peptide nameIL-3 (AF)Protein synonymsMCGF|P-cell stimulating factor|HCGF|Multi-CSFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP01586SequenceMFPTSQVRRG DFTEDTTPNR PVYTTSQVGG LITHVLWEIV EMRKELCNGN SDCMNNDDAL AENNLKLPEI QRNDGCYQTG YNQEICLLKI SSGLLEYHSY LEYMKNNLKD NKKDKARVLQ RDTETLIHIF NQEVKDLHKI VLPTPISNAL LTDKLESQKE WLRTKTIQFI LKSLEEFLKV TLRSTRQTProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight15,2 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-01AF-1MGProtein/peptide nameIL-3 (AF)Protein synonymsMCGF|P-cell stimulating factor|HCGF|Multi-CSFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP01586SequenceMDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVECProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight15,2 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay



