Mouse Recombinant FGF-Basic (from E. coli)
About this item
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
5 Options Available Below
Product Details & Documents
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-12AF-10UG
- SHBT200-12AF-50UG
- SHBT200-12AF-100UG
- SHBT200-12AF-500UG
- SHBT200-12AF-1MG
- Protein/peptide name:
- FGF-Basic (AF)
- FGF-Basic (AF)
- FGF-Basic (AF)
- FGF-Basic (AF)
- FGF-Basic (AF)
- Protein synonyms:
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P15655
- P15655
- P15655
- P15655
- P15655
- Sequence:
- MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- 10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- Molecular weight:
- 16,5 kDa
- 16,5 kDa
- 16,5 kDa
- 16,5 kDa
- 16,5 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-12AF-10UGProtein/peptide nameFGF-Basic (AF)Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP15655SequenceMPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight16,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-12AF-50UGProtein/peptide nameFGF-Basic (AF)Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP15655SequenceMPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight16,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-12AF-100UGProtein/peptide nameFGF-Basic (AF)Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP15655SequenceMPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight16,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-12AF-500UGProtein/peptide nameFGF-Basic (AF)Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP15655SequenceMPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight16,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-12AF-1MGProtein/peptide nameFGF-Basic (AF)Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP15655SequenceMPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5Molecular weight16,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



