Order Entry
Belgium
ContactUsLinkComponent
Rat Recombinant IL-6 (Animal free) (from E. coli)
Rat Recombinant IL-6 (Animal free) (from E. coli)
Certificates
undefined
Rat Recombinant IL-6 (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: B9 cell proliferation, ED50 ≤2 ng/ml, bioactivity ≥5,0×10⁵ units/mg
  • Gene ID:
    P20607
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    IFN-β2|HSF|BSF-2|B-cell differentiation factor|MGI-2
  • UniProtKB:
    P20607
  • Protein/peptide name:
    IL-6
  • Purity:
    ≥90%
  • Molecular weight:
    21,9 kDa
  • Sequence:
    MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA) + 2:1 mannitol to protein
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together