Order Entry
Belgium
ContactUsLinkComponent
 
Dog Recombinant RC GM-CSF (Animal free) (from E. coli)
 
undefined
Dog Recombinant RC GM-CSF (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Dog
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, bioactivity ≥6,7×10⁴ units/mg
  • Gene ID:
    P48749
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    MGI-1GM|CSF-2|Pluripoietin-α
  • UniProtKB:
    P48749
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥95%
  • Molecular weight:
    14,4 kDa
  • Sequence:
    MAPTRSPTLV TRPSQHVDAI QEALSLLNNS NDVTAVMNKA VKVVSEVFDP EGPTCLETRL QLYKEGLQGS LTSLKNPLTM MANHYKQHCP PTPESPCATQ NINFKSFKEN LKDFLFNIPF DCWKPVKK
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together