About this item
Resistin-like molecule-beta (RELM-β) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). Resistin and RELM-β are the only RELM family members found in humans, whereas all four RELM family members are present in rodents. RELM-β functions to increase fibroblast proliferation and differentiation, resulting in airway remodelling and increased inflammation.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Specifications
- Conjugaison:Unconjugated
- Fonction des protéines:Cytokine
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Mouse
- Conditions de stockage:−20 °C
- Teneur en endotoxines:Endotoxin LAL (EU/µg) ≤1
- Activité biologique:Bioactivity assay: B9 cell proliferation, ED50 ≤250 ng/ml, bioactivity ≥4,0×10³ units/mg
- ID de gène:P47873
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Exempt d'endotoxine:N
- Sans vecteur:Y
- Exempt de protéase:N
- Exempt de substance animale:Y
- Synonyme de protéine:oprelvekin|AGIF (Adipogenesis Inhibitory Factor)
- Base de données UnitProt:P47873
- Nom de la protéine/du peptide:IL-11
- Purity:≥95%
- Molecular weight:19,3 kDa
- Sequence:MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL
- Niveau d'endotoxine:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Absence de nucléases:N
- N°:RUO
- Température de transport:RT
- Tested applications:Bioassay