- Description:DECON-QUAT® 200C, ready to mix, SimpleMix
- Seq:06
- Sterility:Non sterile
- Packed:463 ml
- Pk:12 Bottle
DEC-QUAT 200C is a disinfectant, non-food contact sanitizer, cleaner, mildewstat, fungicide, virucide, and deodorizer. DEC-QUAT 200C is a fifth generation quaternary ammonium solution.
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Filtered at 0.2 microns
- One-step disinfectant for use in chemical cleaning rotation cycles
- Phosphate-free
DEC-QUAT 200C is effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, hard surface disinfectant, and non-food contact sanitizer effective against a broad spectrum of bacteria and viruses. DEC-QUAT 200C is a complete, chemically balanced disinfectant/sanitizer that provides clear use solutions even in the presence of hard water.
Applications include hard, inanimate surfaces in filling and gowning rooms, and more generally machinery tables, counter tops, laminar flow benches, floors, and walls, stainless steel, porcelain, glass, and chrome.