Sodium chloride, LiChropur™ for HPLC, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1461 °C (1013 hPa)
Melting Pt:801 °C
Density:2.17 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-598-3
Frequently Bought Together