




TMS-HT (=HMDS and TMCS in anhydrous pyridine) Trimethylsilylating Reagent, for OH compounds
Catalog # TCIAT0274-12ML
Supplier: TCI
CAS Number:
Specifications
- Pk:12 mL
- Seq:2288
Specifications
Specification Test Results
- Appearance:White to Orange to Green clear liquid to cloudy liquid
- Effect test of TMS derivatization:Effective as derivatization agent D-(+)-Glucose
- High boiling impurity of Silylation agent:No impurity higher than Isopropyl myristate(50 ppm) after 5 min + hold up time
Specification Test Results
About this item
:
Flash Pt:20 °C
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
MDL Number:MFCD00192427