Orders LinkContactUsLinkComponent
Labels for thermal transfer printers
Labels for thermal transfer printers
Supplier:   Brady
Labels for thermal transfer printers
Supplier:  Brady
Certificates
Labels for thermal transfer printers
Labels for thermal transfer printers
15 Options Available Below
Product Details & Documents
Thermal transfer printer labels for a variety of uses, including centrifuge, Eppendorf and PCR tube, conical and large tube, and vial laboratory applications.
  • Polypropylene labels with permanent acrylic adhesive
  • Gloss finish
  • Thickness 0,076 mm
  • Maximum service temperature is 120 °C
  • Minimum service temperature is –196 °C
Thermal transfer labels are compatible with Brady printers BBP11, BBP12, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, and BP4320.
Product Family Options
Product Information
  • Model
    Material
    Adhesive
    Printing method
    Colour
    Dimensions
    Packed
    Availability
    Price
  • BRDY302985
    Rectangle and circle printer labels
    Rectangle and circle printer labels
    THT-307-7600-1.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    9,53×25,40 mm
    1500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 144,00 €
    144,00 €
    1 * 1 ROU
       144,00 €
  • BRDY302989
    Rectangle printer labels
    Rectangle printer labels
    THT-301-7600-2.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    9,53×25,40 mm
    2500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 218,00 €
    218,00 €
    1 * 1 ROU
       218,00 €
  • BRDY302990
    Rectangle printer labels
    Rectangle printer labels
    THT-301-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    9,53×25,40 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 216,00 €
    216,00 €
    1 * 1 ROU
       216,00 €
  • BRDY302994
    Rectangle and circle printer labels
    Rectangle and circle printer labels
    THT-304-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    9,53×25,40 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 238,00 €
    238,00 €
    1 * 1 ROU
       238,00 €
  • BRDY302986
    Rectangle and circle printer labels
    Rectangle and circle printer labels
    THT-308-7600-1.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    12,70×25,40 mm
    1500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 157,00 €
    157,00 €
    1 * 1 ROU
       157,00 €
  • BRDY302987
    Rectangle and circle printer labels
    Rectangle and circle printer labels
    THT-309-7600-1.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    12,70×25,40 mm
    1500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 163,00 €
    163,00 €
    1 * 1 ROU
       163,00 €
  • BRDY302984
    Rectangle printer labels
    Rectangle printer labels
    THT-310-7600-2.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    12,70×25,40 mm
    2500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 237,00 €
    237,00 €
    1 * 1 ROU
       237,00 €
  • BRDY302995
    Rectangle and circle printer labels
    Rectangle and circle printer labels
    THT-306-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    12,70×25,40 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 259,00 €
    259,00 €
    1 * 1 ROU
       259,00 €
  • BRDY302991
    Rectangle printer labels
    Rectangle printer labels
    THT-310-7600-10
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    12,70×25,40 mm
    10000 labels/pack
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 725,00 €
    725,00 €
    1 * 10.000 ST
       725,00 €
  • BRDY302982
    Rectangle printer labels
    Rectangle printer labels
    THT-302-7600-1.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    15,24×41,28 mm
    1500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 187,00 €
    187,00 €
    1 * 1 ROU
       187,00 €
  • BRDY302981
    Rectangle printer labels
    Rectangle printer labels
    THT-302-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    15,24×41,28 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 310,00 €
    310,00 €
    1 * 1 ROU
       310,00 €
  • BRDY302988
    Square printer labels
    Square printer labels
    THT-305-7600-2.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    25,40×25,40 mm
    2500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 306,00 €
    306,00 €
    1 * 1 ROU
       306,00 €
  • BRDY302992
    Square printer labels
    Square printer labels
    THT-305-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    25,40×25,40 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 310,00 €
    310,00 €
    1 * 1 ROU
       310,00 €
  • BRDY302983
    Rectangle printer labels
    Rectangle printer labels
    THT-303-7600-1.5SC
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    25,40×48,26 mm
    1500 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 142,00 €
    142,00 €
    1 * 1 ROU
       142,00 €
  • BRDY302993
    Rectangle printer labels
    Rectangle printer labels
    THT-303-7600-3
    PP
    Permanent acrylic
    Thermal transfer
    Clear
    25,40×48,26 mm
    3000 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 238,00 €
    238,00 €
    1 * 1 ROU
       238,00 €