Reference Material for Flash Point Certified by The Japan Petroleum Institute, Hexadecane
TCIAS0555-250ML
: TCI
:
- Pack type:Glass bottle
- Pk:250 mL
- CAS No.:544-76-3
- Description:Reference Material for Flash Point Certified by The Japan Petroleum Institute
- Formula:H₃C(CH₂)₁₄CH₃
- Description:Reference Material for Flash Point Certified by The Japan Petroleum Institute
- Appearance:White powder to lump