Order Entry
Belgium
ContactUsLinkComponent
Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane
Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane
Catalog # TCIAS0554-250ML
Supplier:  TCI
CAS Number:  
undefined
Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane
Catalog # TCIAS0554-250ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • CAS No.:
    124-18-5
  • Description:
    Reference Material for Flash Point Certified by The Japan Petroleum Institute
  • Formula:
    H₃C(CH₂)₈CH₃
  • Description:
    Reference Material for Flash Point Certified by The Japan Petroleum Institute
  • Appearance:
    Colorless clear liquid

Specifications

About this item