Hexane, Standard Material for GC
TCIAS0278-5ML
: TCI
:
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:110-54-3
- Description:Hexane, Purity (GC): min. 99.5 %
- Melting point:"–95 °C (lit.)"
- Formula:H₃C(CH₂)₄CH₃
- Description:Hexane
- Flash point:"–22 °C (lit.)"
- Boiling point:68 °C (lit.)
- Appearance:Colorless clear liquid