Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Food Yellow No. 4
Food Yellow No. 4
Food Yellow No. 4
Catalog # TCIAF0144-25G
Supplier:
TCI
CAS Number:
Expand image
Food Yellow No. 4
Catalog # TCIAF0144-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
CAS No.:
1934-21-0
Description:
Food Yellow No. 4
Formula:
C₁₆H₉N₄Na₃O₉S₂
Description:
Food Yellow No. 4
Specifications
About this item
Product Details
Documentation
Product Details
Documentation