Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Food Yellow No. 4 Aluminum Lake
Food Yellow No. 4 Aluminum Lake
TCIAF0178-25G
:
TCI
:
Food Yellow No. 4 Aluminum Lake
TCIAF0178-25G
:
TCI
:
Pk:
25 g
CAS No.:
12225-21-7
Description:
Food Yellow No. 4 Aluminum Lake
Description:
Food Yellow No. 4 Aluminum Lake