Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Food Red No. 105
Food Red No. 105
Food Red No. 105
TCIAF0001-25G
:
TCI
:
Food Red No. 105
TCIAF0001-25G
:
TCI
:
Pk:
25 g
CAS No.:
632-69-9
Description:
Food Red No. 105
Formula:
C₂₀H₂Cl₄I₄Na₂O₅
Description:
Food Red No. 105