Order Entry
Belgium
ContactUsLinkComponent
Food Blue No. 2
Food Blue No. 2
Catalog # TCIAF0148-25G
Supplier:  TCI
CAS Number:  
undefined
Food Blue No. 2
Catalog # TCIAF0148-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • CAS No.:
    860-22-0
  • Description:
    Food Blue No. 2, Min 85%
  • Formula:
    C₁₆H₈N₂Na₂O₈S₂
  • Description:
    Food Blue No. 2

Specifications

About this item

Food Blue No. 1 consists mainly of disodium 2-[bis[4- [N-ethyl-N-(3-sulfonatophenylmethyl)amino]phenyl]methylio]benzenesulfonate, (C37H34N2Na2O9S3).