The Biosensis Alpha-Synuclein Rapid™ ELISA kit is a sandwich ELISA that allows the quantification of human alpha-synuclein in less than 4 hours in human citrate-plasma, serum and CSF only if used as directed.
- Complete ELISA kit with reagents and detailed protocols
- Fast assay (<4 hours) without compromising accuracy and precision
- Validated for human blood and CSF samples
- Refer to the kit protocol for specific use instructions for blood and CSF application
This ELISA kit consists of a pre-coated sheep polyclonal anti-alpha-synuclein (aa: 116-131) capture antibody, a biotinylated mouse monoclonal anti-alpha-synuclein detection antibody (aa: 61-95) and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3',5,5'-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of alpha-synuclein present in samples and protein standards. A human alpha-synuclein positive control (QC sample) is provided to assure consistent assay performance.
Specificity: Human alpha-synuclein. Detection of individual alpha-synuclein isoforms has not yet been investigated. However, due to the nature of the assay antibodies, this ELISA kit is not expected to discriminate between various alpha-synuclein isoforms (monomeric, oligomeric, phosphorylated).
Cross-reactivity with beta- and gamma-synuclein was calculated based on increase or decrease of apparent endogenous α-synuclein concentrations when spiking beta- and gamma-synuclein into two human plasma samples. This ELISA shows very little cross-reactivity of 2,3% or less for beta- and gamma-synuclein.
- Assay Type:Sandwich
- Format:Pre-coated
- Host:E. coli
- Primary antibody reactivity:Human
- Target protein:alpha-Synuclein (SNCA)
- Storage conditions:2 - 8 °C
- Sample type:Citrate-plasma, serum and CSF
- Detection range:0,16 - 10 ng/ml
- Detection limit:<100 pg/ml