SafeKey lockout padlocks, nylon
About this item
These SafeKey nylon padlocks are the ultimate locks for all types of safety and security needs.
- Lightweight with a ribbed design for easy gripping
- Ideal for electrical applications and enclosures
- Key releases only upon lock closure
- Preferred for most indoor lockout applications
- Engraving options on front, back, left side, right side, and/or key
- Key retaining (Key won’t release until the padlock shackle is securely closed)
- Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout. The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted. The lock mechanism has six precision steel tumbler blades for a high number of key differences and tamper resistance.
Locks are lightweight and resist impact, temperature, chemicals, and corrosion. Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling. The colour-matched key and lock body allows a fast identification in the application.
Informations de commande: Custom keying, engraving, charting, Master Key, and Grand Master Key options are available.
Livraison: One key is supplied with each lock.
63 Options Available Below
Product Details & Documents
These SafeKey nylon padlocks are the ultimate locks for all types of safety and security needs.
- Lightweight with a ribbed design for easy gripping
- Ideal for electrical applications and enclosures
- Key releases only upon lock closure
- Preferred for most indoor lockout applications
- Engraving options on front, back, left side, right side, and/or key
- Key retaining (Key won’t release until the padlock shackle is securely closed)
- Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout. The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted. The lock mechanism has six precision steel tumbler blades for a high number of key differences and tamper resistance.
Locks are lightweight and resist impact, temperature, chemicals, and corrosion. Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling. The colour-matched key and lock body allows a fast identification in the application.
Informations de commande: Custom keying, engraving, charting, Master Key, and Grand Master Key options are available.
Livraison: One key is supplied with each lock.
Documents
Brady SafeKey Lockout Padlocks
Documentation produit
TDS SafeKey Nylon Padlocks TDS SafeKey Nylon Padlocks
Fiche technique
Product Family Options
38 mm steel shackles
Product Information
- ColourW×D×HAvailabilityPrice
- BRDY150234SafeKey padlock, 113,4 g
- BRDY150294SafeKey padlock, 340,2 g
- BRDY150224SafeKey padlock, 680,4 g
- BRDY150251SafeKey padlock, 113,4 g
- BRDY150317SafeKey padlock, 340,2 g
- BRDY150316SafeKey padlock, 680,4 g
- BRDY150275SafeKey padlock, 113,4 g
- BRDY150228SafeKey padlock, 340,2 g
- BRDY150269SafeKey padlock, 680,4 g
- BRDY150368SafeKey padlock, 113,4 g
- BRDY150341SafeKey padlock, 340,2 g
- BRDY150337SafeKey padlock, 680,4 g
- BRDY150320SafeKey padlock, 113,4 g
- BRDY150364SafeKey padlock, 340,2 g
- BRDY150336SafeKey padlock, 680,4 g
- BRDY150250SafeKey padlock, 113,4 g
- BRDY150229SafeKey padlock, 340,2 g
- BRDY150362SafeKey padlock, 680,4 g
- BRDY150321SafeKey padlock, 113,4 g
- BRDY150271SafeKey padlock, 340,2 g
- BRDY150270SafeKey padlock, 680,4 g
- BRDY150367SafeKey padlock, 113,4 g
- BRDY150340SafeKey padlock, 340,2 g
- BRDY150292SafeKey padlock, 680,4 g
- BRDY150343SafeKey padlock, 113,4 g
- BRDY150246SafeKey padlock, 340,2 g
- BRDY150225SafeKey padlock, 680,4 g
76 mm steel shackles
Product Information
- ColourW×D×HAvailabilityPrice
- BRDY150274SafeKey padlock, 149,7 g
- BRDY150249SafeKey padlock, 149,7 g
- BRDY150247SafeKey padlock, 149,7 g
- BRDY150319SafeKey padlock, 149,7 g
- BRDY150248SafeKey padlock, 149,7 g
- BRDY150233SafeKey padlock, 149,7 g
- BRDY150357SafeKey padlock, 149,7 g
- BRDY150295SafeKey padlock, 149,7 g
- BRDY150296SafeKey padlock, 149,7 g
38 mm plastic shackles
Product Information
- ColourW×D×HAvailabilityPrice
- 238-0178SafeKey padlock, 95,3 g
- BRDY150315SafeKey padlock, 285,8 g
- BRDY150351SafeKey padlock, 571,5 g
- 238-0179SafeKey padlock, 95,3 g
- BRDY150268SafeKey padlock, 285,8 g
- BRDY150221SafeKey padlock, 571,5 g
- 238-0180SafeKey padlock, 95,3 g
- BRDY150314SafeKey padlock, 285,8 g
- BRDY150309SafeKey padlock, 571,5 g
- 238-0181SafeKey padlock, 95,3 g
- BRDY150223SafeKey padlock, 285,8 g
- BRDY150334SafeKey padlock, 571,5 g
- 238-0182SafeKey padlock, 95,3 g
- BRDY150222SafeKey padlock, 285,8 g
- BRDY150310SafeKey padlock, 571,5 g
- 238-0183SafeKey padlock, 95,3 g
- BRDY150353SafeKey padlock, 285,8 g
- BRDY150350SafeKey padlock, 571,5 g
- 238-0184SafeKey padlock, 95,3 g
- BRDY150291SafeKey padlock, 285,8 g
- BRDY150311SafeKey padlock, 571,5 g
- 238-0185SafeKey padlock, 95,3 g
- BRDY150267SafeKey padlock, 285,8 g
- BRDY150308SafeKey padlock, 571,5 g
- 238-0186SafeKey padlock, 95,3 g
- BRDY150290SafeKey padlock, 285,8 g
- BRDY150265SafeKey padlock, 571,5 g