Order Entry
Belgium
ContactUsLinkComponent
Corning® Matrigel® Matrix for Organoid Culture, Corning
Corning® Matrigel® Matrix for Organoid Culture, Corning
Catalog # BDAA356255
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® Matrigel® Matrix for Organoid Culture, Corning
Catalog # BDAA356255
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix For Organoid Culture Phenol-Red Free
  • Size:
    10 mL
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford

Specifications

About this item

Corning Matrigel Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.

  • Quality tested for ability to form and maintain stable 3D domes/droplets on multi-well plates
  • Elastic Modulus (matrix stiffness) measured using a rheometer
  • Protein concentrations determined by Lowry method
  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay

Verified to support both mouse and human organoid growth and differentiation from healthy and/or diseased tissue, each lot has a specific elastic modulus value indicating the stiffness of the gel formed and is qualified to form stable 3D domes commonly used in organoid culture protocols. Available in 10 ml LDEV-free, Phenol Red-free format.

Corning Matrigel Matrix for Organoid culture is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.