Saisie de commande
Belgium
ContactUsLinkComponent
Biosafety cabinets, Class II A2, Purifier®
Biosafety cabinets, Class II A2, Purifier®
Supplier:  LABCONCO
   Sustainable Options Available
Certificates
undefined
Biosafety cabinets, Class II A2, Purifier®
Supplier:  LABCONCO
   Sustainable Options Available
Specifications
  • Catalog No:
  • LBCP302381131
  • LBCP302611031
  • LBCP302581131
  • LBCP302611131
  • LBCP302481131
  • LBCP302411131
  • LBCP302380131
  • Environmentally Preferable:
  • Energy Efficient, Sustainable Materials
  • Energy Efficient, Sustainable Materials
  • Energy Efficient, Sustainable Materials
  • Energy Efficient, Sustainable Materials
  • Energy Efficient, Sustainable Materials
  • Energy Efficient, Sustainable Materials
  • Sash opening:
  • 203,2 mm
  • 254 mm
  • 203,2 mm
  • 254 mm
  • 203,2 mm
  • 254 mm
  • 203 mm
  • Exigence électrique:
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz, Schuko
  • 230 V, 50/60 Hz (Schuko)
  • L×P×H int.:
  • 1074,42×792,48×2265,68 - 2418,08 mm
  • 1988,82×792,48×1567,18 mm
  • 1684,02×792,48×2265,68 - 2418,08 mm
  • 1988,82×792,48×2265,68 - 2418,08 mm
  • 1379,22×792,48×2265,68 - 2418,08 mm
  • 1379,22×792,48×2265,68 - 2418,08 mm