

Paraformaldehyde 20% (w/v) in aqueous solution methanol-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30525-89-4
:
Formula:(CH₂O)n
MDL Number:MFCD00133991
ADR:9,III
:ampouled under argon
UN:3334
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance (Color):Clear colorless
- Form:Liquid
- Ash:≤0,01% (Paraformaldehyde content)
- Comment:Manufactured from 95% Paraformaldehyde prills
- Concentration:19,5 to 20% (Formaldehyde)
- Trace Metal:Iron (Fe) ≤2,0% (Paraformaldehyde content)
Specification Test Results
Related Information
Frequently Bought Together